Tisk…
Protein purification spin columns, Vivapure® Ion Exchange (IEX) Mini/Maxi
Dodavatel: Sartorius
VS-IX01DH24 VS-IX01QH24 VS-IX20SH08 VS-IX01SH24 VS-IX20DH08 VS-IX20QH08
516-0125EA 7670 CZK
516-0125 734-0886 734-0892 734-0888 516-0116 734-0890
Protein purification spin columns, Vivapure® Ion Exchange (IEX) Mini/Maxi
Filtry Filtry centrifugační
Vivapure® Ion Exchange (IEX) spin colums are centrifugal devices. They incorporate Sartobind membrane adsorber technology as their chromatography matrix. Sartobind IEX membrane adsorbers are based on stabilised regenerated cellulose and display a microporous structure with a pore size of >3 μm, which is orders of magnitude larger than conventional chromatographic gel materials.
  • Ready to use
  • Replaces time-consuming column chromatography protein purification
  • Scalable to industrial scale applications
  • Available in Mini and Maxi formats
  • Ideal for sample
More Product Information