Tisk…

n-Heptacosane, Supelco®

Dodavatel: Merck

Synonyms: Heptacosane

M-Clarity™ quality level = MQ100

51559-250MG
51559-250MGEA 1870 CZK
51559-250MG
n-Heptacosane, Supelco®
n-Heptacosane
Vzorec: H₃C(CH₂)₂₅CH₃
Bod varu: 442 °C (1013 hPa)
Hustota: 0,7796 g/cm³ (60 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009862
CAS číslo: 593-49-7
EINECS: 209-792-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56441bb504e299076c95042157b8ab1dcb7c3c2e&vid=46695fd1f34ad586f1ed3fc5a6a2122e328e038e&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR