Tisk…
Glass Extraction Thimble, Grade 603G, Whatman™
Dodavatel: Whatman products (Cytiva)
10371005 10371007 10371011 10371019 10371025 10371036 10371042 10371043 10371045 10371055 10371114 2811-308 10371103
515-9005EA 6220 CZK
515-9005 515-9007 533-0012 515-9011 515-9016 515-9025 515-9017 515-9030 515-9031 515-9036 515-9013 WHAT2811-308 537-0027
Glass Extraction Thimble, Grade 603G, Whatman™
Extraktory Patrony extrakční
Cytiva's Whatman 603G borosilicate glass fiber thimbles with inorganic binder are flexibly designed for use in a full range of commercially available Soxhlet apparatus systems.
  • Resistant to temperatures up to 500 °C (932 °F)
  • Manufactured with walls as thin as 1 to 1,5 mm
  • Analysis of hot and acidic gases, including emission control and smoke stack gas monitoring
  • Soxhlet extraction
  • Analysis of pesticide residues
  • Determination of oil/fat content of foods
  • Analysis of oil and grease in solid wastes
Whatman standard glass extraction thimbles are availableMore Product Information