Tisk…
Syringe filters, ReliaPrep™
Dodavatel: Ahlstrom-Munksjö
760526 760502 760506 760514 760516 760517 760523 760501 760530 760528 760513 760543 760503 760504 760507 760509 760510 760512 760515 760518 760519 760520 760522 760524 760529 760531 760532 760534 760535 760536 760537 760538 760539 760540 760541 760542 760548 760549 760511 760525 760527 760508
514-0838EA 25630 CZK
514-0838 514-0814 514-0818 514-0826 514-0828 514-0829 514-0835 514-0813 514-0842 514-0840 514-0825 514-0855 514-0815 514-0816 514-0819 514-0821 514-0822 514-0824 514-0827 514-0830 514-0831 514-0832 514-0834 514-0836 514-0841 514-0843 514-0844 514-0846 514-0847 514-0848 514-0849 514-0850 514-0851 514-0852 514-0853 514-0854 514-0859 514-0860 514-0823 514-0837 514-0839 514-0820
Syringe filters, ReliaPrep™
Filtry Filtry stříkačkové
These syringe filters are suitable for convenient sample preparation and fluid sterilization. The syringe filters for highly loaded aqueous solutions including a glass fiber prefilter and a cellulose acetate membrane are also available with MBS housing and male Luer lock outlet.
  • Transparent housing for full visual control
  • High quality housing construction and welding to prevent leaking
  • Ideal for samples containing proteins and generally recommended for aqueous solutions filtration
  • Nylon, regenerated Cellulose membrane are recommended for organic solvents
More Product Information