Tisk…
Organic vapour monitors
Dodavatel: 3M
3551
514-0007EA 12810 CZK
514-0007
Organic vapour monitors
Systém detekce plynů
Monitor uses the proven scientific principle of diffusion which involves the gradual mixing of molecules of two or more substances for measuring personal exposure in work areas of industries.
  • Ethylene oxide monitor utilises brominated carbon to collect and convert the reactive ethylene oxide to a more stable chemical derivative, 2-bromoethanol
  • Compound then desorbed with methylene chloride in methanol
  • Accuracy within ±25% for 8-hour samplings at 1 ppm
  • Accuracy within ±35% for 8-hour samplings at the action limit
18-month shelf life.
Certifikace: OSHA