Tisk…
Corning® Spin-X® UF Concentrators
Dodavatel: Corning
431477 431478 431479 431480 431481 431482 431483 431484 431485 431486 431489 431487 431488 431490 431491
512-0046EA 3780 CZK
512-0046 512-0047 512-0048 512-0049 512-0050 512-0052 512-0057 512-0058 512-0059 512-0060 512-0063 512-0061 512-0062 512-0080 512-0082
Corning® Spin-X® UF Concentrators
Filtry Filtry centrifugační
These non sterile centrifugal concentrators offer a simple, one step procedure for concentrating or desalting proteins and other biomolecules with 90% or better recovery.
  • Vertical membrane design and thin channel filtration chamber minimises membrane fouling and provides fast, high speed concentrating, even with particle laden solutions
  • Integrated dead stop design eliminates risk of spinning to dryness; no respinning necessary
  • Low binding polyethersulfone (PES) membranes are available with five molecular weight cut-offs (MWCO): 5000, 10000, 30000, 50000
More Product Information