Tisk…
Compressors for syringeless filters, Mini-UniPrep™ G2, Whatman™
Dodavatel: Whatman products (Cytiva)
MUPG2HCPWC1 MUPG2MCPWC8
511-0259EA 668 CZK
511-0259 511-0228
Compressors for syringeless filters, Mini-UniPrep™ G2, Whatman™
Lisy
Whatman™ Mini-UniPrep™ G2 filters from Cytiva can be used with either a hand compressor (single position) or a multicompressor.
  • Mini-UniPrep™ vials can be loaded directly into the autosampler
The Mini-UniPrep™ hand compressor is a time reducing compressor for the Mini-UniPrep™ G2 syringeless filter, a fast and cost-effective syringeless filter that simplifies UHPLC and HPLC sample filtration. Ergonomic hand compressor allows for sample preparation with increased hand comfort. Mini-UniPrep™ all-in-one syringeless filter prepares samples three timesMore Product Information