Tisk…
VWR®, Síta analytická, 200×50 mm
Dodavatel: VWR Collection
510-0700EA 31600 CZK
510-0700 510-0701 510-0702 510-0703 510-0704 510-0705 510-0706 510-0707 510-0708 510-0709 510-0710 510-0711 510-0712 510-0713 510-0714 510-0715 510-0716 510-0717 510-0718 510-0719 510-0720 510-0721 510-0722 510-0723 510-0724 510-0725 510-0726 510-0727 510-0728 510-0729 510-0730 510-0731 510-0732 510-0733 510-0734 510-0735 510-0736 510-0737 510-0738 510-0739 510-0740 510-0741 510-0742 510-0743 510-0744 510-0745 510-0746 510-0747 510-0748 510-0749 510-0750 510-0751 510-0752 510-0753 510-0754 510-0755 510-0756 510-0757 510-0758 510-0759 510-0760 510-0761 510-0762 510-0763 510-0764 510-0765 510-0766 510-0767 510-0768 510-0769 510-0770 510-0771 510-0772 510-0773 510-0774 510-0775 510-0776 510-0777 510-0778 510-0779 510-0780 510-0781 510-0782 510-0783 510-0784 510-0785 510-0786 510-0787 510-0788 510-0789 510-0790 510-0791 510-0792 510-0793 510-0794 510-0795 510-0796 510-0797 510-0798
VWR®, Síta analytická, 200×50 mm
Síta Síta
Rám: Nerezová ocel; tkaná drátěná tkanina: Nerezová ocel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informace oMore Product Information