Tisk…

Perchloric acid 0.01 mol/l (0.01 N) in aqueous solution, Supelco®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

50439-1L
50439-1LEA 1930 CZK
50439-1L
Perchloric acid 0.01 mol/l (0.01 N) in aqueous solution, Supelco®
Perchloric acid
Vzorec: HClO₄
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS číslo: 7601-90-3
EINECS: 231-512-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57680f1dd0f615218c3ca9aeeb229bae28e0d4c5&vid=4bb5b6c6b445d1dd0cdc0721ab4a29c03b23b425&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR