Tisk…

Kyselina sírová 96% ≥96%, SLSI for the electronics industry

Dodavatel: ELECTRONIC CHEMICALS
Nebezpečí

50139211
50139211.EA 0 CZK
50139211.
Kyselina sírová 96% ≥96%, SLSI for the electronics industry
Kyselina sírová

Product from BASF.


Informace o dodávce: This product is not available in all countries. Please check with your local VWR office or supplier.


Upozornení: This product is not available in all countries. Please check with your local VWR office or supplier.

Vzorec: H₂SO₄
Bod varu: ~335 °C (1013 hPa)
Bod tání: 3 °C
Hustota: 1,84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS číslo: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
REACH: 01-2119458838-20
Merck Index: 14,08974

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR