Tisk…

Hydroxid sodný 10 N vodný roztok, Biotech Reagent, J.T.Baker®

Dodavatel: Avantor
Nebezpečí

Synonyms: Caustic soda

5000-35US
5000-03EA 9580 CZK
5000-03 5000-07 5000-02 5000-27 5000-09 5000-39 5000-35 5000-35US
Hydroxid sodný 10 N vodný roztok, Biotech Reagent, J.T.Baker®
Hydroxid sodný

Made from USP purified water and NF (multicompendial) sodium hydroxide pellets that meet Ph. Eur., BP, and JP chemical specifications.
Filtered through a 0.2 µm filter. Completely miscible with water. GMP manufactured product.

Vzorec: NaOH
MW: 40
Bod varu: 119…135 °C (1013 hPa)
Bod tání: 9…10 °C
Hustota: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS číslo: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Objednat

Výsledky specifikačního testu

GMP Manufactured Product Passes Test
IMPORTANT: Material will freeze if stored below 41 °F (5°C). 9.9 - 10.1
Protect from air to avoid absorption of carbon dioxide. = 0.6 %
CAUTION: For Manufacturing, processing or repackaging = 5 ppm
Appearance (Clear, colorless liquid) Passes Test
Normality 9.9 - 10.1
Sodium Carbonate (Na₂CO₃) ≤0.6 %
Chloride (Cl) ≤20 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤2 ppm
Endotoxin Concentration (EU/mL) ≤2.0
Made from U.S.P. Purified Water and Sodium Hydroxide pellets, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
IMPORTANT: Material will freeze if stored below 41°F (5°C).
Filtered through a 0.2 micron filter.

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR