Tisk…
Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: L243]
Dodavatel: Tonbo Biosciences
50-9952-T025 50-9952-T100 50-9952-T500
50-9952-T025EA 1810 CZK
50-9952-T025 50-9952-T100 50-9952-T500
Anti-HLA-DR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: L243]
Protilátky
The L243 antibody reacts with a member of the human MHC Class II antigens, HLA-DR. The HLA-DR antigen is expressed on B lymphocytes, activated T lymphocytes, activated NK cells, monocytes, macrophages, other antigen presenting cells and progenitor cells. The L243 antibody is specific to an epitope on the alpha subunit of the heterodimeric HLA-DR protein and binds a different epitope than the LN3 antibody clone. It does not cross-react with HLA-DP or HLA-DQ. This antibody is reported to be cross-reactive with non-human primates including Chimpanzee, Cynomolgus, Rhesus and Baboon. PleaseMore Product Information