Tisk…
Anti-IL4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11B11]
Dodavatel: Tonbo Biosciences
50-7041-U025 50-7041-U100
50-7041-U025EA 1660 CZK
50-7041-U025 50-7041-U100
Anti-IL4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11B11]
Protilátky
The 11B11 antibody binds to mouse Interleukin-4 (IL-4), a 14 kDa cytokine that is largely secreted by activated T cells of the Th2 subset, and to some degree by NKT and mast cells. This cytokine acts as a stimulatory factor for B cells, inducing their proliferation and differentiation, as well as playing a role in immunoglobulin class-switching. IL-4 may also provide autocrine stimulation for T cells, and affect the function of antigen presenting cells such as macrophages and dendritic cells. IL-4 can bind and signal via three cell surface receptor types: CD124 by itself, CD124 inMore Product Information