Tisk…

Rat IgG2a Isotype Control [clone: 2A3] (PE (Phycoerythrin))

Dodavatel: Tonbo Biosciences
50-4321-U100
50-4321-U100EA 2830 CZK
50-4321-U100
Rat IgG2a Isotype Control [clone: 2A3] (PE (Phycoerythrin))
Protilátky
The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.

Type: Control
Antigen: IgG2a (Rat) (rat IgG2a)
Clonality: monoclonal
Clone: 2A3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR