Tisk…
Anti-CD80 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 16-10A1]
Dodavatel: Tonbo Biosciences
50-0801-U100 50-0801-U025
50-0801-U100EA 3790 CZK
50-0801-U100 50-0801-U025
Anti-CD80 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 16-10A1]
Protilátky
The 16-10A1 antibody reacts with mouse CD80, also known as B7-1, a 55 kDa type I transmembrane protein ligand for CD152 (CTLA-4) and for CD28, a co-stimulatory receptor for the T cell receptor (TCR). CD28 also binds a second B7 ligand known as CD86 (B7-2). Both CD80 and CD86 are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between theMore Product Information
Objednat
Produktová specifikace
Symbol antigenu CD80
Environmentally Preferable
Název antigenu CD80 molecule
Synonyma antigenu TSA1|B71|Ly-53|MIC17|Cluster of Differentiation 80|Cd28l|Ly53
Druh protilátky Primární
Klonovalita Monoclonal
Spojení PE (Phycoerythrin)
Klon 16-10A1
Reaktivita Mouse
Hostitel Armenian hamster
Identifikační číslo genu 12519
Izotyp IgG
Imunochemie Yes
Imunoprecipitace Yes
ImunoFluorescence Yes
Průtoková cytometrie Yes
Křížová adsorbce none
Skladovací teplota Cool
Transportní teplota Ice