Tisk…
Anti-IL2RA Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: PC61.5]
Dodavatel: Tonbo Biosciences
50-0251-U100 50-0251-U025
50-0251-U100EA 3820 CZK
50-0251-U100 50-0251-U025
Anti-IL2RA Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: PC61.5]
Protilátky
The PC61.5 antibody is specific for mouse CD25, a 55 kDa surface protein also known as the Interleukin-2 Receptor alpha chain, or IL-2R alpha. CD25 may bind IL-2 by itself, although with low affinity and without induction of cell signaling. CD25 is also expressed within a high-affinity complex, along with the IL-2R beta chain (CD122) and the common gamma chain (CD132), to form a signaling receptor complex. Expression of CD25 varies during developmental stages of T and B cells, is induced on activated mature T and B cells, and is present on subsets of dendritic cells. CD25 signaling as partMore Product Information
Objednat
Produktová specifikace
Symbol antigenu IL2RA
Environmentally Preferable
Název antigenu Interleukin 2 Receptor, Alpha
Synonyma antigenu TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
Druh protilátky Primární
Klonovalita Monoclonal
Spojení PE (Phycoerythrin)
Klon PC61.5
Reaktivita Mouse
Hostitel Rat
Identifikační číslo genu 16184
Izotyp IgG1 lambda
Imunochemie Yes
Imunoprecipitace Yes
Průtoková cytometrie Yes
Křížová adsorbce none
Skladovací teplota Cool
Transportní teplota Ice