Tisk…

Chlorid měďnatý dihydrát ≥99.0% (by iodometry), crystals, AR® ACS, Macron Fine Chemicals™

Dodavatel: Avantor Performance Materials
Nebezpečí

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

4824-20EA 0 CZK
4824-20
Chlorid měďnatý dihydrát ≥99.0% (by iodometry), crystals, AR® ACS, Macron Fine Chemicals™
Chlorid měďnatý dihydrát
Vzorec: CuCl₂·2H₂O
Bod tání: 100 °C
Hustota: 2,54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS číslo: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8,III
Merck Index: 13,02660

Objednat

Výsledky specifikačního testu

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 99.0 %
ACS - Assay (CuCl₂·2H₂O) (by iodometry) ≥99.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤0.005 %
ACS - Nitrate (NO₃) ≤0.015 %
Solubility Passes Test
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Sodium (Na) ≤0.02 %
ACS - Nickel (Ni) ≤0.01 %
ACS - Potassium (K) ≤0.01 %
ACS - Calcium (Ca) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR