Tisk…
Nunc™ Universal Tubes, Low Profile
Dodavatel: Thermo Fisher Scientific
331840 374320 374321 374322 374323 331840-BR 374321-BR 374320-BR
479-6862EA 2328 CZK
479-6862 479-6863 479-6864 479-6865 479-6866 479-2494 479-2499 479-2498
Nunc™ Universal Tubes, Low Profile
Zkumavky Zkumavky na vzorky
Tubes PP/Rack PC/PP. Low profile 5,0 ml externally-threaded universal tubes provide a robust solution for solid or liquid sample storage. Available in configurations that meet your tracking and packaging requirements, the low profile universal tube features 2D barcodes, graduations, a white patch for labelling, and a wide mouth design for easy addition and extraction of sample material. Its externally-threaded design minimises contamination risk in manual handling procedures. The tubes and rack are rated for storage down to vapor phase liquid nitrogen temperatures.
  • Flexible tube design
More Product Information
Časově omezený prodej
Select items below are currently on sale
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí NAR Certificate of Retirement