Tisk…
VWR®, Cryoboxes, PP, Flat-Packed
Dodavatel: VWR Collection
479-4006EA 2470 CZK
479-4006 479-4001 479-4002 479-4003 479-4004 479-4005
VWR®, Cryoboxes, PP, Flat-Packed
Krabice Kryoboxy
Made of corrugated or thin film PP.
  • Easy self-assembly boxes shipped flat-packed
  • More durable than cardboard boxes as they are not susceptible to moisture
  • Significantly reduces the chance of mould
  • Dividers included with the boxes
  • Store down to temperatures of –80 °C
Designed with sustainability in mind, they are made from either specially formulated corrugated or thin film polypropylene. With superior durability, polypropylene outlasts the cardboard box alternative that is susceptible to softening and producing dust, and can be frozen and thawedMore Product Information