Tisk…
Nunc™ Universal Tubes, Low Profile
Dodavatel: Thermo Fisher Scientific
331840 374320 374321 374322 374323 331840-BR 374321-BR 374320-BR
479-6862EA 2328 CZK
479-6862 479-6863 479-6864 479-6865 479-6866 479-2494 479-2499 479-2498
Nunc™ Universal Tubes, Low Profile
Zkumavky Zkumavky na vzorky
Tubes PP/Rack PC/PP. Low profile 5,0 ml externally-threaded universal tubes provide a robust solution for solid or liquid sample storage. Available in configurations that meet your tracking and packaging requirements, the low profile universal tube features 2D barcodes, graduations, a white patch for labelling, and a wide mouth design for easy addition and extraction of sample material. Its externally-threaded design minimises contamination risk in manual handling procedures. The tubes and rack are rated for storage down to vapor phase liquid nitrogen temperatures.
  • Flexible tube design
More Product Information