Tisk…
Tissue vials, CryoELITE™, WHEATON®
Dodavatel: DWK Life Sciences
W985100
479-2012EA 12690 CZK
479-2012
Tissue vials, CryoELITE™, WHEATON®
Vialky
Designed specifically for tissue collection, transport and storage to provide excellent protection. Different from cells and biofluids, tissue specimens have particular requirements for cryogenic storage. These uniform vials are able to maintain sample integrity while maximising storage capacity and organisation. They feature a wide mouth opening and high integrity closure. Made from low binding, cryogenic grade virgin PP that meets the USP Class VI classification.
  • Designed specifically for tissue sections, suitable for long-term storage in the vapour
More Product Information