Tisk…
Laboratory markers, Nalgene®
Dodavatel: Thermo Fisher Scientific
6310-0010 6311-0010
479-0013EA 792 CZK
479-0013 479-0014
Laboratory markers, Nalgene®
Popisovače Fixy laboratorní
Universal black pens for a wide variety of applications. Cap with handy clip.
  • Special fast drying ink
  • Highly chemical resistant and waterproof when writing on PolyPaper
  • Also suitable for marking glass and plastics