Tisk…
Dewar vessels, cylindrical, type C and G-C
Dodavatel: KGW
1011 10111 10112 10114 10115 10117 10118 1012 10120 10122 10123 10124 1013 1014 1015 1016 1018 1019 1021 10211 10212 10214 10215 10217 10218 1022 10220 10222 10223 10224 1023 1024 1025 1026 1028 1029 1061 10610 10611 10612 10613 10614 1062 1063 1064 1065 1066 1067 1068 1069 10616 10615 10213 10216 10221 10210 1027 10113
478-0714EA 1910 CZK
478-0714 478-0214 478-0715 478-0717 478-0718 478-0719 478-0720 478-0721 478-0722 478-0723 478-0724 478-0725 478-0726 478-0727 478-5147 478-0728 478-0729 478-0730 478-0215 478-4110 478-4114 478-4115 478-4120 478-4116 478-4121 478-4102 478-4125 478-4130 478-4131 478-4140 478-4103 478-0764 478-4105 478-4109 478-4108 478-0216 478-0217 478-4141 478-0218 478-0219 478-0220 KGWA10614 KGWA1062 478-0221 478-0222 478-0731 478-0223 478-0224 478-0225 KGWA1069 KGWA10616 KGWA10615 478-0763 KGWA10216 KGWA10221 KGWA10210 KGWA1027 478-0716
Dewar vessels, cylindrical, type C and G-C
Dewarovy a kryogenní nádoby
Dewar vessel, in silvered borosilicate glass, blue coated metal protective covering. For storage of liquid nitrogen, dry ice and other coolants.
Objednat
Produktová specifikace
Model 00 A 9 A 10 A 12 A 13 A 15 A 16 A 0 A 18 A 20 A 21 A 22 A 11 A 2 A 3 A 4 A 6 A 7 A 00 C 9 C 10 C 12 C 13 C 15 C 16 C 0 C 18 C 20 C S 21 C S 22 C 1 C 2 C 3 C 4 C 6 C 7 C G 0 C G 12 C G 13 C G 15 C G 16 C G 18 C G 1 C G 2 C G 3 C G 4 C G 6 C G 7 C G 9 C G 10 C GS 21 C G 20 C 11 C 14 C 19 C 8 C 5 C 11 A
Kapacita 0,1 L 1 L 1,5 L 1,5 L 2 L 1,5 L 2 L 0,2 L 2,5 L 3 L 4 L 8 L 0,3 L 0,45 L 0,5 L 0,75 L 0,8 L 1,2 L 0,1 L 1 L 1,5 L 1,5 L 2 L 1,5 L 2 L 0,2 L 2,5 L 3 L 4 L 8 L 0,3 L 0,45 L 0,5 L 0,75 L 0,8 L 1,2 L 0,2 L 1,5 L 2 L 1,5 L 2 L 2,5 L 0,3 L 0,45 L 0,5 L 0,75 L 0,8 L 1,2 L 1 L 1,5 L 4 L 3 L 2,1 L 3,2 L 5 L 1,7 L 1,25 L 2,1 L
Environmentally Preferable
Rozsah teplot −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200 ...+150 °C
Denní vypařování 0,3 L 0,35 L 0,4 L 0,5 L 0,5 L 0,7 L 0,6 L 0,2 L 0,7 L 1,2 L 1,0 L 0,9 L 0,2 L 0,25 L 0,35 L 0,3 L 0,35 L 0,3 L 0,2 L 0,5 L 0,5 L 0,7 L 0,6 L 0,7 L 0,2 L 0,25 L 0,35 L 0,3 L 0,35 L 0,3 L 0,35 L 0,4 L 1,0 L 1,2 L 0,4 L 0,45 L 0,6 L 0,3 L 0,25 L
Int.Ø 40 mm 77 mm 77 mm 90 mm 90 mm 100 mm 100 mm 40 mm 110 mm 138 mm 138 mm 138 mm 47 mm 47 mm 57 mm 57 mm 67 mm 67 mm 40 mm 77 mm 77 mm 90 mm 90 mm 100 mm 100 mm 40 mm 110 mm 138 mm 138 mm 138 mm 47 mm 47 mm 57 mm 57 mm 67 mm 67 mm 40 mm 90 mm 90 mm 100 mm 100 mm 110 mm 57 mm 47 mm 57 mm 57 mm 67 mm 67 mm 77 mm 77 mm 138 mm 138 mm 77 mm 90 mm 110 mm 67 mm 57 mm 77 mm
Int. height 90 mm 235 mm 345 mm 245 mm 340 mm 240 mm 290 mm 170 mm 290 mm 230 mm 310 mm 600 mm 190 mm 270 mm 210 mm 310 mm 240 mm 350 mm 90 mm 235 mm 345 mm 245 mm 340 mm 240 mm 290 mm 170 mm 290 mm 230 mm 310 mm 600 mm 190 mm 270 mm 210 mm 310 mm 240 mm 350 mm 170 mm 245 mm 340 mm 240 mm 290 mm 290 mm 210 mm 270 mm 210 mm 310 mm 240 mm 350 mm 235 mm 345 mm 310 mm 230 mm 130 mm 600 mm 600 mm 500 mm 500 mm 500 mm
Overall height 135 mm 290 mm 395 mm 300 mm 390 mm 300 mm 345 mm 215 mm 345 mm 280 mm 375 mm 660 mm 240 mm 310 mm 260 mm 370 mm 295 mm 395 mm 215 mm 300 mm 390 mm 300 mm 345 mm 345 mm 240 mm 310 mm 260 mm 370 mm 295 mm 395 mm 290 mm 395 mm 375 mm 280 mm 550 mm 665 mm 660 mm 560 mm 550 mm