Tisk…

Platina nominally 20% on carbon black

Dodavatel: Thermo Fisher Scientific

Synonyms: Platinium

47381.06 47381.03
47381.06EA 18330 CZK
47381.06 47381.03
Platina nominally 20% on carbon black
Platina
Vzorec: Pt
MDL Number: MFCD00011179
CAS číslo: 7440-06-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR