Tisk…

Paraformaldehyd 8% (w/v) vodný roztok methanol-free

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Ampuled under argon

47347.9L 47347.9M
47347.9LEA 1490 CZK
47347.9L 47347.9M
Paraformaldehyd 8% (w/v) vodný roztok methanol-free
Paraformaldehyd

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent.


Solution is micro-filtered and supplied in ampoules. RNase and DNase free.

Vzorec: (CH₂O)n
Storage Temperature: Ambient
MDL Number: MFCD00133991
CAS číslo: 30525-89-4

Objednat

Výsledky specifikačního testu

Appearance (Color) Clear colorless
Form Liquid
Ash ≤0.01% (Paraformaldehyde content)
Comment Starting material RNase and DNase free
Concentration 7.5 to 8% (Formaldehyde)
Trace Metal Iron (Fe) ≤2.0% (Paraformaldehyde content)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR