Tisk…
Immersion coolers, FT Series
Dodavatel: JULABO GmbH
9650820 9650890 9650840 9650842 9650892 9650893
473-3005EA 84720 CZK
473-3005 473-3020 473-3306 473-0019 473-0020 JULA9650893
Immersion coolers, FT Series
Termostaty oběhové Termostaty oběhové Termostat cirkulační s chlazením
FT immersion coolers are primarily used for counter cooling when connected to a heating circulator, or for rapidly cooling fluids down to low temperatures. The compact, easy to use units are suitable for a variety of cooling applications.
  • Efficient cooling with reduced energy consumption
  • Easy to move around the laboratory by convenient carrying handles, larger FT900/902 have castors
  • Alternative to cooling with dry ice or tap water
  • FT402, FT902 and FT903 provide temperature control via a keypad and have an external PT100 sensor
  • Environmentally friendly as
More Product Information