Tisk…

Tellurium, plasma standard solution, Te 1000 µg/ml, Specpure®

Dodavatel: Thermo Fisher Scientific

47183.AEEA 3350 CZK
47183.AE 47183.AD
Tellurium, plasma standard solution, Te 1000 µg/ml, Specpure®
Standardy Standardy kationické Standardy - tellur

Molecular Formula: Te in 30% HCl
Physical Form: Liquid
Odour: Acidic
UN No.: UN1789
MDL No.: MFCD00134062

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR