Tisk…

Indole-3-butyric acid potassium salt 98%

Dodavatel: Thermo Fisher Scientific

471050010 471050050 471050250
471050010.EA 1120 CZK
471050010. 471050050. 471050250.
Indole-3-butyric acid potassium salt 98%
Indole-3-butyric acid potassium salt
Vzorec: C₁₂H₁₂KNO₂
Storage Temperature: Ambient
MDL Number: MFCD00058442
CAS číslo: 60096-23-3

Objednat

Výsledky specifikačního testu

Appearance white to yellow crystalline powder
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %
Water ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR