Tisk…
Laboratory Fridge-Freezer with SmartFrost, Perfection SCFfg
Dodavatel: Liebherr
SCFFG4002
471-1342EA 62490 CZK
471-1342
Laboratory Fridge-Freezer with SmartFrost, Perfection SCFfg
Chladničky
Liebherr appliances are optimised for maximum energy efficiency and low operating costs. We only use the natural and environmentally-friendly refrigerants R 600a. When used with highly efficient compressors, these are extraordinarily efficient and, with their low global warming potential, also future-proof.
  • ±5 K temperature stability for freezers
  • ±3 K temperature stability for Refrigerators
  • Mechanical lock
  • Antibacterial lever handle
  • Replaceable door seal
  • For training purposes or exhibitions, the functionalities of the appliance can be simulated in demo
More Product Information