Tisk…

Sucralose 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside

470090010 470091000 470092500
470090010.EA 5920 CZK
470090010. 470091000. 470092500.
Sucralose 98%
Sucralose
Vzorec: C₁₂H₁₉Cl₃O₈
Storage Temperature: Ambient
MDL Number: MFCD03648615
CAS číslo: 56038-13-2
EINECS: 259-952-2
Merck Index: 13,08965

Objednat

Výsledky specifikačního testu

Appearance white to almost white crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +84° to +88° (20°C, 589 nm) (C=1% in water) (on dry substance)
Water ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR