Tisk…

4-Isoxazolecarboxylic acid 98%

Dodavatel: Thermo Scientific

Synonyms: Isoxazole-4-carboxylic acid

469640010
469640010.EA 4310 CZK
469640010.
4-Isoxazolecarboxylic acid 98%
4-Isoxazolecarboxylic acid
Vzorec: C₄H₃NO₃
MW: 113,0715 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06738806
CAS číslo: 6436-62-0

Objednat

Výsledky specifikačního testu

Appearance light yellow to pink to brown powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 120°C to 125°C
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR