Tisk…

Molybden ≥99.95% (metals basis), foil, Thickness: 0.05 mm (0.002 in)

Dodavatel: Thermo Fisher Scientific

46943.GH 46943.HW 46943.HX
46943.GHEA 2920 CZK
46943.GH 46943.HW 46943.HX
Molybden ≥99.95% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Molybden
Vzorec: Mo
MW: 95.94
Bod varu: 3617
Bod tání: 2622
Hustota: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS číslo: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR