Tisk…
Desiccator plates, Nalgene®
Dodavatel: Thermo Fisher Scientific
5312-0230
467-3100EA 3800 CZK
467-3100
Desiccator plates, Nalgene®
Exsikátory Desky do exsikátorů
Unbreakable desiccator plates with fire polished, corrosion resistant, inert, non stick glass surface bonded to metal. Suitable for use with desiccators: 467-3120, 467-3110 and 467-3112.
  • Greater resistance to thermal shock than porcelain
  • Marked with numbered quadrants for easy reference
  • Autoclavable