Tisk…
Water baths, PURA™
Dodavatel: JULABO GmbH
9550504 9550510 9550514 9550522 9550530
462-0625EA 26090 CZK
462-0625 462-0626 462-0627 462-0628 462-0629
Water baths, PURA™
Lázně Lázně vodní, nemíchané
PURA stainless steel water baths are functional and reliable for everyday operations due to their high material and component quality. The easy to use PURA water baths offer a number of functions and features that enable efficient use in the laboratory.
  • Intuitive, straightforward menu and bright, easy to read LED display
  • Splash-proof protected power switch
  • Integral drain screw (except PURA 4) and integrated timer
  • Built-in dry running protection
  • No interfering elements in the bath
  • Optical and acoustic alarm
  • Non slip rubber feet and ergonomic recessed grips
More Product Information