Tisk…
Compatible control refrigerated circulators
Dodavatel: HUBER
2019.0001.01 2023.0001.01 2022.0001.01 2021.0001.01 2020.0002.01 2025.0001.01 2017.0001.01 2017.0002.01 2018.0001.01 2018.0002.01 2024.0001.01 2025.0002.01 2027.0001.01 2027.0002.01 2026.0005.01 2036.0001.01 2019.0004.01 2020.0010.01 2021.0005.01 2044.0001.01 2044.0002.01 2045.0001.01 2045.0004.01
461-1063EA 476420 CZK
461-1063 461-1068 461-6463 461-0620 461-1065 461-1069 461-6460 461-1060 461-6461 461-1061 461-0567 461-1070 461-1072 461-1073 461-1071 461-1074 461-1064 461-1067 HUBE2021.0005.01 HUBE2044.0001.01 HUBE2044.0002.01 HUBE2045.0001.01 HUBE2045.0004.01
Compatible control refrigerated circulators
Termostaty oběhové Termostaty oběhové Termostat cirkulační s chlazením
These compact refrigerated circulators with insulated refrigeration baths are ideal for direct temperature control from −90 up to +200 °C. The low bath volume and high heating and cooling capacity means short heating and cooling rates, which can be increased by using the optional displacement inserts. Optional calibration inserts enable circulators to be used as calibration baths.
  • Pilot ONE® is interchangeable and can be used as a remote control (with an optional data cable)
  • Controller can be upgraded with enhanced functionality at any time (Exclusive and
More Product Information