Tisk…
Crucibles, porcelain
Dodavatel: MORGAN ADVANCED MATERIALS HALD
L79/0 L79/00 L79/000A L79/1 L79/2 L79/2A L79/3 L79/4 L79C/0 L79C/00 L79C/1 L79C/2 L79C/3 L79C/4 L79C/5 L79MF/1 L79MF/1A L79MF/2 L79MF/3 L79MF/4 L79MF/5 L79MF/6 L79MF/6A L79MF/7 L79MF/7A L79MF/8 L79MF/9
459-0101EA 221 CZK
459-0101 459-0102 459-0103 459-0104 459-0136 459-0105 459-0137 410-9325 459-1122 459-1121 459-0138 459-1120 459-0139 459-0140 459-0141 459-1110 459-1118 459-0142 459-0143 459-0144 459-0145 459-0146 459-0147 459-0106 459-0148 459-1111 459-0149
Crucibles, porcelain
Kelímky
Porcelain, glazed inside and out, except base.
  • Withstand maximum temperature of 1000 °C
Lids are available separately.