Tisk…

Silikagel, Pore size: 120 Å ≥99.99%, spherical 7.5 µm APS for preparative chromatography

Dodavatel: Thermo Fisher Scientific
Varování

45816.14 45816.06
45816.14EA 7420 CZK
45816.14 45816.06
Silikagel, Pore size: 120 Å ≥99.99%, spherical 7.5 µm APS for preparative chromatography
Silikagel

Purity: 99.99+% CAS No.: 63231-67-4 Physical Form: S.A. 335m2/g, P.V. 1.00cc/g MDL No.: MFCD00011232

Vzorec: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS číslo: 63231-67-4
EINECS: 231-545-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR