Tisk…
zinek 1.000 mg/l v zřed. kyselině dusičné (from Zn) ARISTAR® standard pro ICP
Dodavatel: VWR Chemicals
456384KEA 7980 CZK
456384K 456382Y
zinek 1.000 mg/l v zřed. kyselině dusičné (from Zn) ARISTAR® standard pro ICP
Standardy Standardy kationické Standardy - zinek
Zn in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.