Tisk…
VWR® ARISTAR®, Antimony Standard Solutions for ICP
Dodavatel: VWR Chemicals
455022GEA 3580 CZK
455022G 455024Y 455034K
VWR® ARISTAR®, Antimony Standard Solutions for ICP
Standardy Standardy kationické Standardy - antimon
Sb in HNO₃ 2% tr. tartaric acid.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.
Certifikace: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Informace o dodávce: Supplied with certificate of analysis.