Tisk…

Silver(I) citrate hydrate

Dodavatel: Thermo Fisher Scientific
Varování

45496.06 45496.14 45496.22
45496.06EA 3610 CZK
45496.06 45496.14 45496.22
Silver(I) citrate hydrate
Silver(I) citrate hydrate

CAS No.: 206986-90-5 Molecular Formula: Ag3C6H5O7.xH2O Formula Weight: 512.70(anhy) Physical Form: Powder Sensitivity: Light Sensitive Melting Point: 170° dec. MDL No.: MFCD00150589

Vzorec: C₆H₅Ag₃O₇·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00150589
CAS číslo: 206986-90-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR