Tisk…

Molybden ≥99.9% (metals basis), wire, hard, Ø 0.203 mm (0.008 in)

Dodavatel: Thermo Fisher Scientific

45134.G6 45134.G5 45134.G1
45134.G6EA 7620 CZK
45134.G6 45134.G5 45134.G1
Molybden ≥99.9% (metals basis), wire, hard, Ø 0.203 mm (0.008 in)
Molybden

Purity: 99.9% CAS No.: 7439-98-7 Physical Form: ≈0.33g/m MDL No.: MFCD00003465

Vzorec: Mo
Bod varu: 3617 °C (1013 hPa)
Bod tání: 2622 °C
Hustota: 10,28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS číslo: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR