Tisk…
Spirit lamp, DURAN®
Dodavatel: DWK Life Sciences
294020007 294030008
451-3111EA 27.6 CZK
451-3111 451-3112
Spirit lamp, DURAN®
Ohřívače Kahany
Soda-lime glass with ground glass joint cap. The spirit lamp is an alternative to the gas-powered Bunsen burner and is also suitable for heating cooking glasses and the materials they contain.
Capacity: 100 ml.
Ø×H: 75×103 mm.
Informace pro objednávání: Supplied without socket and wick.