Tisk…
Infrared heaters, IRB series
Dodavatel: Edmund Bühler GmbH
6063000 6060000 9066000
451-1502EA 19430 CZK
451-1502 451-1500 451-1505
Infrared heaters, IRB series
Ohřívače Desky topné
The IRB infrared heaters heated up samples to 700 or 900 °C within a short time.
  • Ideal for use where unshielded flames are not allowed for safety reasons
Objednat
Produktová specifikace
Environmentally Preferable
Model IRB 2 IRB 1 IRB 6
Dimensions 150×150×150 mm 100×105×105 mm 697×305×122 mm
Weight 2,5 kg 0,7 kg 9 kg