Tisk…

Kyselina chlorovodíková 37%, ARISTAR® for trace analysis

Dodavatel: VWR Chemicals
Nebezpečí

Synonyms: Hydrochloric acid solution, Muriatic acid

450021YEA 4610 CZK
450021Y 450027X
Kyselina chlorovodíková 37%, ARISTAR® for trace analysis
Kyselina chlorovodíková
Vzorec: HCl
MW: 36.46 g/mol
Bod tání: –30 °C
Hustota: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS číslo: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
REACH: 01-2119484862-27

Objednat

Výsledky specifikačního testu

Assay 36.0 - 38.0 %
Colour Max. 10 APHA
Br (Bromide) Max. 50 ppm
Free chlorine Max. 0.5 ppm
PO4 (Phosphate) Max. 0.1 ppm
SO4 (Sulphate) Max. 0.5 ppm
SO3 (Sulphite) Max. 0.500 ppm
Ag (Silver) Max. 0.010 ppm
Al (Aluminium) Max. 0.050 ppm
As (Arsenic) Max. 0.010 ppm
Au (Gold) Max. 0.020 ppm
B (Boron) Max. 0.010 ppm
Ba (Barium) Max. 0.005 ppm
Be (Beryllium) Max. 0.010 ppm
Bi (Bismuth) Max. 0.020 ppm
Ca (Calcium) Max. 0.100 ppm
Cd (Cadmium) Max. 0.003 ppm
Co (Cobalt) Max. 0.005 ppm
Cr (Chromium) Max. 0.005 ppm
Cu (Copper) Max. 0.002 ppm
Fe (Iron) Max. 0.050 ppm
Ga (Gallium) Max. 0.020 ppm
Hg (Mercury) Max. 0.001 ppm
In (Indium) Max. 0.010 ppm
K (Potassium) Max. 0.010 ppm
Li (Lithium) Max. 0.010 ppm
Mg (Magnesium) Max. 0.020 ppm
Mn (Manganese) Max. 0.005 ppm
Mo (Molybdenum) Max. 0.005 ppm
NH4 (Ammonium) Max. 1 ppm
Na (Sodium) Max. 0.050 ppm
Ni (Nickel) Max. 0.010 ppm
Pb (Lead) Max. 0.002 ppm
Pt (Platinum) Max. 0.010 ppm
Sn (Tin) Max. 0.010 ppm
Sr (Strontium) Max. 0.005 ppm
Ti (Titanium) Max. 0.010 ppm
Tl (Thallium) Max. 0.010 ppm
V (Vanadium) Max. 0.0050 ppm
Zn (Zinc) Max. 0.010 ppm
Zr (Zirconium) Max. 0.005 ppm
Ignition residue (SO4) Max. 2.0 ppm
Residue on evaporation Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR