Tisk…

Lead selenide ≥99.999% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

44916.22 44916.14
44916.22EA 32710 CZK
44916.22 44916.14
Lead selenide ≥99.999% (metals basis)
Lead selenide

Purity: 99.999+% CAS No.: 12069-00-0 Molecular Formula: PbSe Formula Weight: 286.15 Physical Form: 3-6mm Fused Lump Odour: Product specific UN No.: 3077 MDL No.: MFCD00016273

Vzorec: PbSe
Hustota: 8,1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016273
CAS číslo: 12069-00-0
EINECS: 235-109-4
UN: 3077
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR