Tisk…

Zlato ≥99.96% (metals basis), spherical powder APS 0,5-0,8 µm

Dodavatel: Thermo Fisher Scientific

44636.03 44636.06 44636.MD
44636.03EA 9850 CZK
44636.03 44636.06 44636.MD
Zlato ≥99.96% (metals basis), spherical powder APS 0,5-0,8 µm
Zlato

Purity: 99.96+% CAS No.: 7440-57-5 Physical Form: S.A. 0.45-0.70m2/g MDL No.: MFCD00003436

Vzorec: Au
Bod varu: 2800 °C (1013 hPa)
Bod tání: 1063 °C
Hustota: 19,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS číslo: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR