Tisk…

2-Chlorotrityl chloride 1,0 - 1,4 mmol/g on polystyrene, cross-linked with 1% DVB 100-200 mesh

Dodavatel: Thermo Fisher Scientific

Synonyms: (2-Chlorophenyl)diphenylmethyl chloride, Chloro(2-chlorophenyl)diphenylmethane, (Chloro(2-chlorophenyl)methylene)dibenzene

44407.04 44407.09 44407.18
44407.04EA 3510 CZK
44407.04 44407.09 44407.18
2-Chlorotrityl chloride 1,0 - 1,4 mmol/g on polystyrene, cross-linked with 1% DVB 100-200 mesh
2-Chlorotrityl chloride

CAS No.: 42074-68-0 Physical Form: Powder MDL No.: MFCD00040399

Vzorec: C₁₉H₁₄Cl₂
Storage Temperature: Ambient
MDL Number: MFCD00040399
CAS číslo: 42074-68-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR