Tisk…

Tris[N,N-bis(trimethylsilyl)amide]gadolinium(III) ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

44359.03 44359.06 AB204444.0001
44359.03EA 3920 CZK
44359.03 44359.06 ABCRAB204444.0001
Tris[N,N-bis(trimethylsilyl)amide]gadolinium(III) ≥98%
Tris[N,N-bis(trimethylsilyl)amide]gadolinium(III)

Purity: 98% CAS No.: 35789-03-8 Molecular Formula: C18H54GdN3Si6 Formula Weight: 638.41 Physical Form: Powder Sensitivity: Moisture Sensitive UN No.: 3396 MDL No.: MFCD03427091

Vzorec: C₁₈H₅₄GdN₃Si₆
Bod tání: 161…167 °C
Storage Temperature: Ambient
MDL Number: MFCD03427091
CAS číslo: 35789-03-8
UN: 3396
ADR: 4.3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR