Tisk…

n-Oktadekan ≥90%, tech.

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Octadecane

44301.A1 44301.A7
44301.A1EA 2610 CZK
44301.A1 44301.A7
n-Oktadekan ≥90%, tech.
n-Oktadekan

Purity: 90% CAS No.: 593-45-3 Molecular Formula: CH3(CH2)16CH3 Formula Weight: 254.50 Flash Point: 166°(330°F) Physical Form: Crystalline mass/melt Density: 0.782 Refractive Index: 1.4390 Appearance: White Odour: Odourless Melting Point: 28° Boiling Point: 317° MDL No.: MFCD00009007

Vzorec: H₃C(CH₂)₁₆CH₃
MW: 254.5
Bod varu: 317 °C
Bod tání: 28 °C
Hustota: 0.7765
Bod vzplanutí: 166 °C (330 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009007
CAS číslo: 593-45-3
EINECS: 209-790-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR