Tisk…
Magnetic stirring bars, tapered
Dodavatel: COWIE
001.1915 001.1920 001.1925 001.1930 001.1935 001.1940 001.1950 001.1960 001.1970 001.1980 001.1910 001.1945
442-9615EA 240 CZK
442-9615 442-9620 442-9625 442-9630 442-0066 442-9640 442-9650 442-9660 442-9670 442-9680 442-9610 442-0067
Magnetic stirring bars, tapered
Míchadla Míchadla magnetická
PTFE covered, strong Alnico V magnetic core. Smooth surface. Stirring action is similar to oval or pivot ring type stirring bars.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)